Product Details

SNP ID
rs1602914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5286813 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACAACAAACGTAAGCAAATGATA[C/T]GGGTTGTTAGATGATAAGTGCAATG
Phenotype
MIM: 603616
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RABEP1 PubMed Links

Gene Details

Gene
RABEP1
Gene Name
rabaptin, RAB GTPase binding effector protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083585.2 Intron NP_001077054.1
NM_001291581.1 Intron NP_001278510.1
NM_001291582.1 Intron NP_001278511.1
NM_004703.5 Intron NP_004694.2
XM_011524060.1 Intron XP_011522362.1
XM_017025294.1 Intron XP_016880783.1
XM_017025295.1 Intron XP_016880784.1

View Full Product Details