Product Details

SNP ID
rs11803138
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:88694275 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTATTCTCTGTGTTCTACCTATT[C/T]ATCCTTCCCTCTCCTCCATCCCCTG
Phenotype
MIM: 602549
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKN2 PubMed Links

Gene Details

Gene
PKN2
Gene Name
protein kinase N2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320707.1 Intron NP_001307636.1
NM_001320708.1 Intron NP_001307637.1
NM_001320709.1 Intron NP_001307638.1
NM_006256.3 Intron NP_006247.1
XM_011541772.2 Intron XP_011540074.1
XM_017001782.1 Intron XP_016857271.1
XM_017001783.1 Intron XP_016857272.1
Gene
PKN2-AS1
Gene Name
PKN2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details