Product Details

SNP ID
rs35803911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112568405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATAAAGTTTATGTTTATCCTTGG[A/C]AAAATTGTAAAGCGAGTTGTTTGTA
Phenotype
MIM: 609606
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATG3 PubMed Links

Gene Details

Gene
ATG3
Gene Name
autophagy related 3
There are no transcripts associated with this gene.

Gene
SLC35A5
Gene Name
solute carrier family 35 member A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017945.3 Intron NP_060415.1
XM_011512948.1 Intron XP_011511250.1

View Full Product Details