Product Details
- SNP ID
-
rs2307028
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52808596 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGAGTCTGTTCATATCTGACTTCTA[C/T]TGGGCTTGAGCTAATGATCACCTGT
- Phenotype
-
MIM: 123940
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT4
PubMed Links
Gene Details
- Gene
- KRT4
- Gene Name
- keratin 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002272.3 |
|
Intron |
|
|
NP_002263.3 |
View Full Product Details