Product Details

SNP ID
rs787206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120328533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATTAGACTTCGCTAATTCTGT[C/T]TTTTAAAAACCCCAATTAAAGTTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC58 PubMed Links

Gene Details

Gene
LRRC58
Gene Name
leucine rich repeat containing 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099678.1 3879 UTR 3 NP_001093148.1

View Full Product Details