Product Details

SNP ID
rs8077539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4846641 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAGTAATAAGTGATTGAAGTGTT[C/T]GTGTTTAAAGCAGAGCCTCTATGGG
Phenotype
MIM: 609426
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MINK1 PubMed Links

Gene Details

Gene
MINK1
Gene Name
misshapen like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024937.3 Intron NP_001020108.1
NM_001321236.1 Intron NP_001308165.1
NM_015716.4 Intron NP_056531.1
NM_153827.4 Intron NP_722549.2
NM_170663.4 Intron NP_733763.1
XM_006721531.2 Intron XP_006721594.1
XM_006721532.2 Intron XP_006721595.1
XM_006721536.2 Intron XP_006721599.1
XM_011523906.1 Intron XP_011522208.1
XM_017024704.1 Intron XP_016880193.1
XM_017024705.1 Intron XP_016880194.1
XM_017024706.1 Intron XP_016880195.1
XM_017024707.1 Intron XP_016880196.1
XM_017024708.1 Intron XP_016880197.1

View Full Product Details