Product Details

SNP ID
rs12931
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:72974156 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCGCCCACCTTCCGAGCAATGAG[C/T]GTTCCACCTTCATCGTCCACTTGTG
Phenotype
MIM: 605471
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZFYVE1 PubMed Links

Gene Details

Gene
ZFYVE1
Gene Name
zinc finger FYVE-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281734.1 882 Silent Mutation NP_001268663.1
NM_001281735.1 882 Silent Mutation NP_001268664.1
NM_021260.3 882 Silent Mutation NP_067083.1
NM_178441.2 882 Silent Mutation NP_848535.1
XM_017021373.1 882 Silent Mutation XP_016876862.1

View Full Product Details