Product Details

SNP ID
rs1048604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169083707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGAATACAGTCTGTTATTTACTT[A/C]TCTCTTTTAACATAAGGTTGGGAAC
Phenotype
MIM: 165215
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MECOM PubMed Links

Gene Details

Gene
MECOM
Gene Name
MDS1 and EVI1 complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105077.3 4623 UTR 3 NP_001098547.3
NM_001105078.3 4623 UTR 3 NP_001098548.2
NM_001163999.1 4623 UTR 3 NP_001157471.1
NM_001164000.1 4623 UTR 3 NP_001157472.1
NM_001205194.1 4623 UTR 3 NP_001192123.1
NM_004991.3 4623 UTR 3 NP_004982.2
NM_005241.3 4623 UTR 3 NP_005232.2
XM_005247213.3 4623 UTR 3 XP_005247270.1
XM_005247214.3 4623 UTR 3 XP_005247271.1
XM_005247215.3 4623 UTR 3 XP_005247272.1
XM_005247219.2 4623 UTR 3 XP_005247276.1
XM_005247220.2 4623 UTR 3 XP_005247277.1
XM_005247221.2 4623 UTR 3 XP_005247278.1
XM_005247223.2 4623 UTR 3 XP_005247280.1
XM_005247224.3 4623 UTR 3 XP_005247281.1
XM_005247225.3 4623 UTR 3 XP_005247282.1
XM_005247226.3 4623 UTR 3 XP_005247283.1
XM_011512546.2 4623 UTR 3 XP_011510848.1
XM_017005874.1 4623 UTR 3 XP_016861363.1
XM_017005875.1 4623 Intron XP_016861364.1
XM_017005876.1 4623 UTR 3 XP_016861365.1
XM_017005877.1 4623 UTR 3 XP_016861366.1
XM_017005878.1 4623 Intron XP_016861367.1

View Full Product Details