Product Details

SNP ID
rs6504949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:54972772 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCAGCAGAGACCCTAGAAAATTT[G/T]TGTGTATAAACAAGTGTGCATATGT
Phenotype
MIM: 603648 MIM: 610415
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COX11 PubMed Links

Gene Details

Gene
COX11
Gene Name
COX11, cytochrome c oxidase copper chaperone
There are no transcripts associated with this gene.

Gene
STXBP4
Gene Name
syntaxin binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178509.5 Intron NP_848604.3
XM_005257187.4 Intron XP_005257244.1
XM_006721797.3 Intron XP_006721860.1
XM_006721798.3 Intron XP_006721861.1
XM_017024410.1 Intron XP_016879899.1
XM_017024411.1 Intron XP_016879900.1
XM_017024412.1 Intron XP_016879901.1
XM_017024413.1 Intron XP_016879902.1
XM_017024414.1 Intron XP_016879903.1
XM_017024415.1 Intron XP_016879904.1

View Full Product Details