Product Details

SNP ID
rs9929731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:53599340 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACCATGATAATTTAGAAAACATG[C/G]TGAAACTGAGAGTGAAGAGAAATGT
Phenotype
MIM: 610937
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPGRIP1L PubMed Links

Gene Details

Gene
RPGRIP1L
Gene Name
RPGRIP1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005255867.1 Intron XP_005255924.1
XM_005255868.1 Intron XP_005255925.1
XM_011522970.1 Intron XP_011521272.1
XM_011522971.2 Intron XP_011521273.1
XM_011522973.2 Intron XP_011521275.1
XM_017023094.1 Intron XP_016878583.1
XM_017023095.1 Intron XP_016878584.1
XM_017023096.1 Intron XP_016878585.1
XM_017023097.1 Intron XP_016878586.1
XM_017023098.1 Intron XP_016878587.1
XM_017023099.1 Intron XP_016878588.1
XM_017023100.1 Intron XP_016878589.1

View Full Product Details