Product Details
- SNP ID
-
rs34096625
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
30
- Location
-
Chr.1:108858479 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCGTCAGGGATTCATGAATTTT[A/G]TATACAGTGTATGATGTTTCTAGAT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKNAD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148870238] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKNAD1
- Gene Name
- AKNA domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152763.4 |
|
Intron |
|
|
NP_689976.2 |
- Gene
- SPATA42
- Gene Name
- spermatogenesis associated 42 (non-protein coding)
There are no transcripts associated with this gene.
View Full Product Details