Product Details

SNP ID
rs11602674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43365933 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTACCATATGTATGCTGAGAGTA[C/T]CTGCATTCCTTTTGTAACTTAGTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTC17 PubMed Links

Gene Details

Gene
TTC17
Gene Name
tetratricopeptide repeat domain 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307943.1 Intron NP_001294872.1
NM_018259.5 Intron NP_060729.2
XM_005253016.1 Intron XP_005253073.1
XM_006718263.1 Intron XP_006718326.1
XM_011520218.2 Intron XP_011518520.1
XM_017018010.1 Intron XP_016873499.1

View Full Product Details