Product Details

SNP ID
rs2302765
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:7447656 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCATGAAGGGACTTTCATTGGTGAG[C/T]AGGCATGGCTCCTACATCCATGGGC
Phenotype
MIM: 100710 MIM: 601514 MIM: 613936
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRNB1 PubMed Links

Gene Details

Gene
CHRNB1
Gene Name
cholinergic receptor nicotinic beta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000747.2 Intron NP_000738.2
Gene
FGF11
Gene Name
fibroblast growth factor 11
There are no transcripts associated with this gene.

Gene
TMEM102
Gene Name
transmembrane protein 102
There are no transcripts associated with this gene.

View Full Product Details