Product Details

SNP ID
rs2839198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46288340 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGCAACTTGGCATTTCAAACACA[C/T]GTATCCTTTAGTCCTGTGGTGACAC
Phenotype
MIM: 603294
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCM3AP PubMed Links

Gene Details

Gene
MCM3AP
Gene Name
minichromosome maintenance complex component 3 associated protein
There are no transcripts associated with this gene.

Gene
YBEY
Gene Name
ybeY metallopeptidase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006114.2 Intron NP_001006114.1
NM_001314022.1 Intron NP_001300951.1
NM_001314023.1 Intron NP_001300952.1
NM_001314024.1 Intron NP_001300953.1
NM_001314025.1 Intron NP_001300954.1
NM_001314026.1 Intron NP_001300955.1
NM_058181.2 Intron NP_478061.1
XM_005261156.3 Intron XP_005261213.2
XM_011529629.2 Intron XP_011527931.1
XM_011529630.2 Intron XP_011527932.1
XM_011529631.2 Intron XP_011527933.1
XM_011529633.2 Intron XP_011527935.1
XM_011529634.2 Intron XP_011527936.2
XM_011529635.2 Intron XP_011527937.1
XM_017028394.1 Intron XP_016883883.1
XM_017028395.1 Intron XP_016883884.1
XM_017028396.1 Intron XP_016883885.1

View Full Product Details