Product Details

SNP ID
rs11761426
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:100359270 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGTCCCCAATCTAGAAAGCAGCAC[A/T]CCATGCCTTTCATCCAGACGCCCCA
Phenotype
MIM: 605342
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
MIR6840 PubMed Links

Gene Details

Gene
MIR6840
Gene Name
microRNA 6840
There are no transcripts associated with this gene.

Gene
PILRB
Gene Name
paired immunoglobin-like type 2 receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178238.3 Intron NP_839956.1
Gene
PVRIG2P
Gene Name
poliovirus receptor related immunoglobulin domain containing 2, pseudogene
There are no transcripts associated with this gene.

Gene
STAG3L5P-PVRIG2P-PILRB
Gene Name
STAG3L5P-PVRIG2P-PILRB readthrough
There are no transcripts associated with this gene.

View Full Product Details