Product Details

SNP ID
rs1064281
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:6103075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCAGCCCAAAGACGCCAAACATA[C/T]GGACACCACCATCACCACTTTTGGC
Phenotype
MIM: 610771 MIM: 601142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHD5 PubMed Links

Gene Details

Gene
CHD5
Gene Name
chromodomain helicase DNA binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015557.2 8375 UTR 3 NP_056372.1
Gene
KCNAB2
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 2
There are no transcripts associated with this gene.

View Full Product Details