Product Details

SNP ID
rs1004041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1221890 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAAGGAAGAGGCCGGGGAGGAA[G/T]AAGCCAGCCAGGAGGGGGAGCCTGG
Phenotype
MIM: 607904 MIM: 191081 MIM: 609341
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CACNA1H PubMed Links

Gene Details

Gene
CACNA1H
Gene Name
calcium voltage-gated channel subunit alpha1 H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005407.1 1605 Intron NP_001005407.1
NM_021098.2 1605 Intron NP_066921.2
XM_005255652.4 1605 Intron XP_005255709.1
XM_006720963.3 1605 Intron XP_006721026.1
XM_006720964.3 1605 Intron XP_006721027.1
XM_006720965.3 1605 Intron XP_006721028.1
XM_006720967.3 1605 Intron XP_006721030.1
XM_006720968.3 1605 Intron XP_006721031.1
XM_011522724.2 1605 Intron XP_011521026.1
XM_011522727.2 1605 Intron XP_011521029.1
XM_017023819.1 1605 Intron XP_016879308.1
XM_017023820.1 1605 Intron XP_016879309.1
XM_017023821.1 1605 Intron XP_016879310.1
Gene
TPSB2
Gene Name
tryptase beta 2 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
TPSG1
Gene Name
tryptase gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012467.3 1605 Missense Mutation TTA,TTC L288F NP_036599.3
XM_011522446.2 1605 Missense Mutation TTA,TTC L313F XP_011520748.1
XM_011522447.2 1605 Missense Mutation TTA,TTC L313F XP_011520749.1

View Full Product Details