Product Details

SNP ID
rs3087918
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:100831626 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATGCCTTCTCTCACAGGGCTCTGCG[G/T]CTCCGGTTCACTTCTGTGTCTTTAC
Phenotype
MIM: 605636
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MEG3 PubMed Links

Gene Details

Gene
MEG3
Gene Name
maternally expressed 3 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details