Product Details

SNP ID
rs2295283
Assay Type
Validated
NCBI dbSNP Submissions
98
Location
Chr.1:12022869 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CTTCTCTGAGGAGTCTGCAGTTCCT[A/G]AGAGGAGCTGGCGCCTCAGGCCATA
Phenotype
MIM: 608507 MIM: 608772
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFN2 PubMed Links

Gene Details

Gene
MFN2
Gene Name
mitofusin 2
There are no transcripts associated with this gene.

Gene
MIIP
Gene Name
migration and invasion inhibitory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021933.3 616 Missense Mutation AAG,GAG K167E NP_068752.2
XM_005263487.3 616 Missense Mutation AAG,GAG K167E XP_005263544.1
XM_011541895.1 616 Missense Mutation AAG,GAG K167E XP_011540197.1
XM_011541896.1 616 Missense Mutation AAG,GAG K167E XP_011540198.1
Gene
MIR6729
Gene Name
microRNA 6729
There are no transcripts associated with this gene.

View Full Product Details