Product Details
- SNP ID
-
rs2269269
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
27
- Location
-
Chr.1:63601617 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- TGTGTTAAGTAAGTAATGGCTAGCC[A/G]AGGGCTTCAGTTGGGAGGACTGCGG
- Phenotype
-
MIM: 171900
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PGM1
PubMed Links
Gene Details
- Gene
- PGM1
- Gene Name
- phosphoglucomutase 1
View Full Product Details