Product Details

SNP ID
rs7715401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:44305748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGGGGGATGTCTTTCTTCCAAA[C/T]ATCAAATTTCTTAATTTAATGAAAT
Phenotype
MIM: 602115
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF10 PubMed Links

Gene Details

Gene
FGF10
Gene Name
fibroblast growth factor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004465.1 Intron NP_004456.1
XM_005248264.3 Intron XP_005248321.1

View Full Product Details