Product Details
- SNP ID
-
rs2759009
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128836729 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCCTGGGTTCTAACTACCAGGCCTG[C/T]GCTGACTCTCAGGTGGGGTCAAGGC
- Phenotype
-
MIM: 600547
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KYAT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72758869] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KYAT1
- Gene Name
- kynurenine aminotransferase 1
- Gene
- SPOUT1
- Gene Name
- SPOUT domain containing methyltransferase 1
There are no transcripts associated with this gene.
View Full Product Details