Product Details

SNP ID
rs642498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102686674 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAGCTCCCCCACCTCACCTCACC[C/T]GTGGGATTGACAACAGAGATTCAGA
Phenotype
MIM: 615259 MIM: 190470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
METTL21C PubMed Links

Gene Details

Gene
METTL21C
Gene Name
methyltransferase like 21C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010977.2 Intron NP_001010977.1
XM_017020407.1 Intron XP_016875896.1
XM_017020408.1 Intron XP_016875897.1
Gene
TPP2
Gene Name
tripeptidyl peptidase 2
There are no transcripts associated with this gene.

View Full Product Details