Product Details

SNP ID
rs564577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39435417 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGCAAACCCTTAATCTTCAACA[A/C]CCCCATCTCAGCTTTTACATCTTCT
Phenotype
MIM: 611893 MIM: 603675
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC723805 PubMed Links

Gene Details

Gene
LOC723805
Gene Name
interleukin-like
There are no transcripts associated with this gene.

Gene
PLEKHG2
Gene Name
pleckstrin homology and RhoGEF domain containing G2
There are no transcripts associated with this gene.

Gene
RPS16
Gene Name
ribosomal protein S16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001020.5 Intron NP_001011.1
NM_001321111.1 Intron NP_001308040.1
XM_005259137.2 Intron XP_005259194.1

View Full Product Details