Product Details
- SNP ID
-
rs7144509
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:52556889 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CAACATATTGAAGTTCTTACTTTCA[A/G]TTTGGTCTTAGGAGTGCCAGTTGCT
- Phenotype
-
MIM: 616179
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GPR137C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75699608] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPR137C
- Gene Name
- G protein-coupled receptor 137C
- Gene
- TXNDC16
- Gene Name
- thioredoxin domain containing 16
There are no transcripts associated with this gene.
View Full Product Details