Product Details

SNP ID
rs34351158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28668361 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTCTTTATTCTAGAGTAGTTCCC[A/G]CTTGTCAGTCTCAGACTGCAGTGCT
Phenotype
MIM: 602934 MIM: 601333
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDF2 PubMed Links

Gene Details

Gene
SDF2
Gene Name
stromal cell derived factor 2
There are no transcripts associated with this gene.

Gene
SUPT6H
Gene Name
SPT6 homolog, histone chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320755.1 Intron NP_001307684.1
NM_003170.4 Intron NP_003161.2
XM_017024978.1 Intron XP_016880467.1
XM_017024979.1 Intron XP_016880468.1
XM_017024980.1 Intron XP_016880469.1

View Full Product Details