Product Details

SNP ID
rs34167533
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:35491469 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGTTATGCCCGTCCTGCTGTATGGC[C/T]AGCCACTGTTTCTCTTCTGTTTTGA
Phenotype
MIM: 611053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8B5P PubMed Links

Gene Details

Gene
ATP8B5P
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
There are no transcripts associated with this gene.

Gene
RUSC2
Gene Name
RUN and SH3 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135999.1 Intron NP_001129471.1
XM_006716896.2 Intron XP_006716959.1

View Full Product Details