Product Details

SNP ID
rs3807191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:37909196 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGACAAAAGTCTTAAATTATTATA[A/C]ATACAGGAAAAGATTCTGCTGTGTA
Phenotype
MIM: 607421 MIM: 606570
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NME8 PubMed Links

Gene Details

Gene
NME8
Gene Name
NME/NM23 family member 8
There are no transcripts associated with this gene.

Gene
SFRP4
Gene Name
secreted frizzled related protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003014.3 Intron NP_003005.2

View Full Product Details