Product Details
- SNP ID
-
rs7769948
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:157716982 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAATCCCATGCCCGTCTGTCATCTC[C/T]GCACTTGCTGGGCTGCATTTTACTG
- Phenotype
-
MIM: 605952
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SNX9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111627075] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNX9
- Gene Name
- sorting nexin 9
View Full Product Details