Product Details

SNP ID
rs9808136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38877383 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAATTTCACATCTTGCAGACATGT[A/C]AATAACTAATTAATGAGCACACATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DHX57 PubMed Links

Gene Details

Gene
DHX57
Gene Name
DEAH-box helicase 57
There are no transcripts associated with this gene.

Gene
MORN2
Gene Name
MORN repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145450.1 Intron NP_001138922.1

View Full Product Details