Product Details
- SNP ID
-
rs1293987
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
53
- Location
-
Chr.1:186296816 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAGTTAGTGGTGAGATGAAAGAGC[A/T]GTTTTCTGATACTTTTATTTTATTT
- Phenotype
-
MIM: 604283
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRG4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79536913] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRG4
- Gene Name
- proteoglycan 4
View Full Product Details