Product Details

SNP ID
rs4871998
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:22450637 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACGCTGTCATCGTGATCTGTGAAC[A/T]ATATCAAGGGTCATTCTTGTGCTCA
Phenotype
MIM: 114107
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP3CC PubMed Links

Gene Details

Gene
PPP3CC
Gene Name
protein phosphatase 3 catalytic subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243974.1 Intron NP_001230903.1
NM_001243975.1 Intron NP_001230904.1
NM_005605.4 Intron NP_005596.2
XM_017013610.1 Intron XP_016869099.1
XM_017013611.1 Intron XP_016869100.1

View Full Product Details