Product Details

SNP ID
rs11083708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43404463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCCCTCTTTGGAAAGGGATAGCA[C/T]TTTAATATATAATATACTACAATTT
Phenotype
MIM: 612665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEX101 PubMed Links

Gene Details

Gene
TEX101
Gene Name
testis expressed 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130011.1 Intron NP_001123483.1
NM_031451.4 Intron NP_113639.4
XM_005259303.3 Intron XP_005259360.2

View Full Product Details