Product Details
- SNP ID
-
rs4734020
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:101055599 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGTTTATGCATTTAAAAAGAATAT[C/G]TTCTATTATATACATCTAAAAAGTT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ42969
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115461314] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLJ42969
- Gene Name
- uncharacterized LOC441374
There are no transcripts associated with this gene.
View Full Product Details