Product Details
- SNP ID
-
rs678862
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:11544598 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTAGCCAGTTTTAGAACTCCTACC[A/C]AGTTCAGCCTGCTATTTATTTCCCC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM170B
PubMed Links
Gene Details
- Gene
- TMEM170B
- Gene Name
- transmembrane protein 170B
View Full Product Details