Product Details

SNP ID
rs536141
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:117233820 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACAGGCAGACACCTCCAAAAACTTC[A/C]CTTAAATTAATGGAGAATGCTTTGA
Phenotype
MIM: 604872
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PCSK7 PubMed Links

Gene Details

Gene
PCSK7
Gene Name
proprotein convertase subtilisin/kexin type 7
There are no transcripts associated with this gene.

Gene
RNF214
Gene Name
ring finger protein 214
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077239.1 Intron NP_001070707.1
NM_001278249.1 Intron NP_001265178.1
NM_207343.3 Intron NP_997226.2

View Full Product Details