Product Details

SNP ID
rs12805708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:128902448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTGCACAGGGTGCCAGTTAGGG[A/C]GGAGAAGGCAGCGAGGAACCCCGCA
Phenotype
MIM: 600734
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C11orf45 PubMed Links

Gene Details

Gene
C11orf45
Gene Name
chromosome 11 open reading frame 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256088.1 741 UTR 3 NP_001243017.1
NM_145013.2 741 UTR 3 NP_659450.1
XM_011542654.2 741 UTR 3 XP_011540956.1
XM_011542655.2 741 UTR 3 XP_011540957.1
Gene
KCNJ5
Gene Name
potassium voltage-gated channel subfamily J member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000890.3 741 Intron NP_000881.3
XM_011542809.2 741 Intron XP_011541111.1
XM_011542810.2 741 Intron XP_011541112.1

View Full Product Details