Product Details

SNP ID
rs1256148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64466637 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACTTTATCTTTCTGTTAGAGGCA[C/T]ACAGTATGAAACACCCAACCTAAGA
Phenotype
MIM: 604688 MIM: 172460 MIM: 194541
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAP5 PubMed Links

Gene Details

Gene
AKAP5
Gene Name
A-kinase anchoring protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004857.3 Intron NP_004848.3
Gene
MTHFD1
Gene Name
methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1
There are no transcripts associated with this gene.

Gene
ZBTB25
Gene Name
zinc finger and BTB domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304507.1 Intron NP_001291436.1
NM_001304508.1 Intron NP_001291437.1
NM_006977.3 Intron NP_008908.2
XM_005268051.3 Intron XP_005268108.1
XM_006720250.3 Intron XP_006720313.1
XM_006720251.3 Intron XP_006720314.1
XM_006720252.3 Intron XP_006720315.1
XM_011537139.1 Intron XP_011535441.1
XM_011537141.2 Intron XP_011535443.1
XM_011537143.2 Intron XP_011535445.1
XM_011537146.2 Intron XP_011535448.1
XM_017021630.1 Intron XP_016877119.1
XM_017021631.1 Intron XP_016877120.1
XM_017021632.1 Intron XP_016877121.1
XM_017021633.1 Intron XP_016877122.1
XM_017021634.1 Intron XP_016877123.1
XM_017021635.1 Intron XP_016877124.1
XM_017021636.1 Intron XP_016877125.1
XM_017021637.1 Intron XP_016877126.1
XM_017021638.1 Intron XP_016877127.1

View Full Product Details