Product Details

SNP ID
rs11655584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39982771 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATACAGAAGTGCCTAAGAAACTA[C/T]TCATTGATTGCATTCCCTTATTGCT
Phenotype
MIM: 611218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSDMA PubMed Links

Gene Details

Gene
GSDMA
Gene Name
gasdermin A
There are no transcripts associated with this gene.

Gene
PSMD3
Gene Name
proteasome 26S subunit, non-ATPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002809.3 Intron NP_002800.2

View Full Product Details