Product Details

SNP ID
rs4795406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39943881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGAGGAGAAAGCTGGCAGTGGG[C/G]CATGCGATTCTCTTGACTCACTCCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC3C PubMed Links

Gene Details

Gene
LRRC3C
Gene Name
leucine rich repeat containing 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195545.1 Intron NP_001182474.1
XM_011524158.2 Intron XP_011522460.1
XM_017024003.1 Intron XP_016879492.1

View Full Product Details