Product Details

SNP ID
rs12814932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64188639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTGGAGGAAAATAAATAATCA[A/C]CAGTAATTAAGAGAGAAAATTAACC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C12orf66 PubMed Links

Gene Details

Gene
C12orf66
Gene Name
chromosome 12 open reading frame 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300940.1 Intron NP_001287869.1
NM_001300941.1 Intron NP_001287870.1
NM_152440.4 Intron NP_689653.3

View Full Product Details