Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001146310.1 | 644 | Intron | NP_001139782.1 | ||
NM_001256945.1 | 644 | Intron | NP_001243874.1 | ||
NM_001256946.1 | 644 | Intron | NP_001243875.1 | ||
NM_001256947.1 | 644 | Intron | NP_001243876.1 | ||
NM_001282670.1 | 644 | Intron | NP_001269599.1 | ||
NM_001282671.1 | 644 | Silent Mutation | GCA,GCG | A31A | NP_001269600.1 |
NM_001282672.1 | 644 | Silent Mutation | GCA,GCG | A31A | NP_001269601.1 |
NM_001282673.1 | 644 | Intron | NP_001269602.1 | ||
NM_182533.2 | 644 | Intron | NP_872339.2 | ||
XM_006710419.3 | 644 | Intron | XP_006710482.1 | ||
XM_006710421.3 | 644 | Intron | XP_006710484.1 | ||
XM_011540914.2 | 644 | Intron | XP_011539216.1 | ||
XM_011540921.2 | 644 | Intron | XP_011539223.1 | ||
XM_011540922.1 | 644 | Intron | XP_011539224.1 | ||
XM_017000553.1 | 644 | Intron | XP_016856042.1 | ||
XM_017000554.1 | 644 | Intron | XP_016856043.1 | ||
XM_017000555.1 | 644 | Silent Mutation | GCA,GCG | A31A | XP_016856044.1 |
XM_017000556.1 | 644 | Silent Mutation | GCA,GCG | A31A | XP_016856045.1 |
XM_017000557.1 | 644 | Intron | XP_016856046.1 |