Product Details

SNP ID
rs35807494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114123761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACTTAAGTGTCTATCTTAAACA[C/T]TGTAAAAAATCCAACAAGATAGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC186 PubMed Links

Gene Details

Gene
CCDC186
Gene Name
coiled-coil domain containing 186
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321829.1 3689 UTR 3 NP_001308758.1
NM_018017.3 3689 UTR 3 NP_060487.2
NM_153249.1 3689 Intron NP_694981.1
XM_011539915.2 3689 UTR 3 XP_011538217.1
XM_011539916.2 3689 Intron XP_011538218.1

View Full Product Details