Product Details

SNP ID
rs12554914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86028812 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGAAGAAGAAAGCCATACCCCC[A/G]TTGCATGCCCTGTGAAGGGGATAAG
Phenotype
MIM: 606804
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLM1 PubMed Links

Gene Details

Gene
GOLM1
Gene Name
golgi membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016548.3 Intron NP_057632.2
NM_177937.2 Intron NP_808800.1
Gene
NAA35
Gene Name
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
There are no transcripts associated with this gene.

View Full Product Details