Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256279.1 | 848 | Intron | NP_001243208.1 | ||
NM_001256280.1 | 848 | Intron | NP_001243209.1 | ||
NM_019591.3 | 848 | Intron | NP_062537.2 | ||
XM_005266182.3 | 848 | Intron | XP_005266239.1 | ||
XM_005266183.3 | 848 | Intron | XP_005266240.1 | ||
XM_011534829.1 | 848 | Intron | XP_011533131.1 | ||
XM_017019920.1 | 848 | UTR 5 | XP_016875409.1 | ||
XM_017019921.1 | 848 | Intron | XP_016875410.1 | ||
XM_017019922.1 | 848 | Intron | XP_016875411.1 | ||
XM_017019923.1 | 848 | Intron | XP_016875412.1 |