Product Details

SNP ID
rs3818761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124858666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTTTCCATCCTTGGCTCCCACT[C/G]CTGCCCCACTTGTTCTCCAGCAGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARPC5L PubMed Links

Gene Details

Gene
ARPC5L
Gene Name
actin related protein 2/3 complex subunit 5 like
There are no transcripts associated with this gene.

Gene
RPL35
Gene Name
ribosomal protein L35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007209.3 Intron NP_009140.1
Gene
WDR38
Gene Name
WD repeat domain 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045476.2 Intron NP_001038941.1
NM_001276374.1 Intron NP_001263303.1
NM_001276375.1 Intron NP_001263304.1
NM_001276376.1 Intron NP_001263305.1
XM_005251987.3 Intron XP_005252044.1

View Full Product Details