Product Details
- SNP ID
-
rs2270678
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:20387623 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGTCTGCGAAGTTTCCGGAGCCC[C/G]GGTCCCGCCGCGGGTTCGCGCTTGT
- Phenotype
-
MIM: 605628
MIM: 603190
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HTATIP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78245135] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HTATIP2
- Gene Name
- HIV-1 Tat interactive protein 2
There are no transcripts associated with this gene.
- Gene
- PRMT3
- Gene Name
- protein arginine methyltransferase 3
View Full Product Details