Product Details

SNP ID
rs34225151
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5620681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGGTTATGGGTGCACATTATG[A/C]CTTACGGTGGGAGGGGCAGAAGTAC
Phenotype
MIM: 605684 MIM: 608487 MIM: 607564
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
TRIM34 PubMed Links
Additional Information
For this assay, SNP(s) [rs111537348] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TRIM34
Gene Name
tripartite motif containing 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003827.1 Intron NP_001003827.1
NM_021616.5 Intron NP_067629.2
NM_130390.2 Intron NP_569074.2
Gene
TRIM5
Gene Name
tripartite motif containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033034.2 Intron NP_149023.2
NM_033092.2 Intron NP_149083.2
NM_033093.2 Intron NP_149084.2
XM_005253183.2 Intron XP_005253240.1
XM_005253184.2 Intron XP_005253241.1
XM_006718358.2 Intron XP_006718421.1
XM_011520426.2 Intron XP_011518728.1
XM_011520427.1 Intron XP_011518729.1
XM_017018460.1 Intron XP_016873949.1
XM_017018461.1 Intron XP_016873950.1
XM_017018462.1 Intron XP_016873951.1
XM_017018463.1 Intron XP_016873952.1
Gene
TRIM6
Gene Name
tripartite motif containing 6
There are no transcripts associated with this gene.

Gene
TRIM6-TRIM34
Gene Name
TRIM6-TRIM34 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003819.3 Intron NP_001003819.1

View Full Product Details