Product Details

SNP ID
rs10818476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120809760 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCCCAGCTGTGTAACCTTGGGCA[A/G]TTTTCTTAACCTCTCTGTGCAGTCA
Phenotype
MIM: 604452
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMD5 PubMed Links

Gene Details

Gene
PSMD5
Gene Name
proteasome 26S subunit, non-ATPase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270427.1 Intron NP_001257356.1
NM_005047.3 Intron NP_005038.1
XM_011518866.2 Intron XP_011517168.1
XM_017014927.1 Intron XP_016870416.1
XM_017014928.1 Intron XP_016870417.1

View Full Product Details