Product Details
- SNP ID
-
rs12870643
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:112653355 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAAGAGAAAGAAAATGAGAAAAC[C/G]AGGAAGAGCAAGAAAAGGAGGAACA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATP11AUN
PubMed Links
Gene Details
- Gene
- ATP11AUN
- Gene Name
- ATP11A upstream neighbor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207440.2 |
|
Intron |
|
|
NP_997323.1 |
View Full Product Details